Anti-TMEM38B Rabbit Polyclonal Antibody

Supplier: Proteintech
19919-1-AP
10156-034EA 635.16 CAD
10156-034
Anti-TMEM38B Rabbit Polyclonal Antibody
Antibodies
The TMEM38B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TMEM38B. This antibody recognizes human antigen. The TMEM38B antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

WB 1 : 300

Type: Primary
Antigen: TMEM38B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR