Anti-COQ4 Rabbit Polyclonal Antibody

Supplier: Proteintech
16654-1-AP
10154-694EA 818.44 CAD
10154-694
Anti-COQ4 Rabbit Polyclonal Antibody
Antibodies
The COQ4 Antibody from Proteintech is a rabbit polyclonal Antibody to a recombinant protein of human COQ4. This Antibody recognizes human,mouse,rat Antigen. The COQ4 Antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; IF 1 : 50 ; WB 1 : 600

Type: Primary
Antigen: COQ4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR