Anti-VIPR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14878-1-AP
10154-588EA 635.16 CAD
10154-588
Anti-VIPR1 Rabbit Polyclonal Antibody
Antibodies
The VIPR1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human VIPR1. This antibody recognizes human antigen. The VIPR1 antibody has been validated for the following applications: ELISA, IHC analysis.

IF 1 : 200 ; WB 1 : 300

Type: Primary
Antigen: VIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR