Anti-HSCB Rabbit Polyclonal Antibody

Supplier: Proteintech
15132-1-AP
10154-598EA 635.16 CAD
10154-598
Anti-HSCB Rabbit Polyclonal Antibody
Antibodies
The HSCB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HSCB. This antibody recognizes human antigen. The HSCB antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 2000

Type: Primary
Antigen: HSCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR