Anti-C10ORF58 Rabbit Polyclonal Antibody

Supplier: Proteintech
20446-1-AP
10156-056EA 635.16 CAD
10156-056
Anti-C10ORF58 Rabbit Polyclonal Antibody
Antibodies
The C10orf58 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C10orf58. This antibody recognizes human, rat antigen. The C10orf58 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 800

Type: Primary
Antigen: C10ORF58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR