Anti-NDUFB5 Rabbit Polyclonal Antibody

Supplier: Proteintech
23855-1-AP-150UL
10156-086EA 635.16 CAD
10156-086
Anti-NDUFB5 Rabbit Polyclonal Antibody
Antibodies
The NDUFB5 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human NDUFB5. This antibody recognizes human, mouse antigen. The NDUFB5 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300

Type: Primary
Antigen: NDUFB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR