Anti-HERPUD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20640-1-AP
10154-882EA 635.16 CAD
10154-882
Anti-HERPUD2 Rabbit Polyclonal Antibody
Antibodies
The HERPUD2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human HERPUD2. This antibody recognizes human antigen. The HERPUD2 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

WB 1 : 300

Type: Primary
Antigen: HERPUD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR