Anti-SMO Rabbit Polyclonal Antibody

Supplier: Proteintech
20787-1-AP
10154-890EA 635.16 CAD
10154-890
Anti-SMO Rabbit Polyclonal Antibody
Antibodies
The SMO antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SMO. This antibody recognizes human antigen. The SMO antibody has been validated for the following applications: ELISA, IHC analysis.

LL

Type: Primary
Antigen: SMO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR