Anti-ACBD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20941-1-AP
10154-906EA 635.16 CAD
10154-906
Anti-ACBD4 Rabbit Polyclonal Antibody
Antibodies
The ACBD4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ACBD4. This antibody recognizes human antigen. The ACBD4 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 300

Type: Primary
Antigen: ACBD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR