Anti-VAMP8 Rabbit Polyclonal Antibody

Supplier: Proteintech
15546-1-AP
10154-614EA 635.16 CAD
10154-614
Anti-VAMP8 Rabbit Polyclonal Antibody
Antibodies
The VAMP8 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human VAMP8. This antibody recognizes human,mouse,rat antigen. The VAMP8 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1200 ; IP 1 : 700

Type: Primary
Antigen: VAMP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR