Anti-CTDSP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10952-1-AP
10154-420EA 635.16 CAD
10154-420
Anti-CTDSP1 Rabbit Polyclonal Antibody
Antibodies
The CTDSP1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CTDSP1. This antibody recognizes human,mouse,rat antigen. The CTDSP1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 800

Type: Primary
Antigen: CTDSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR