Anti-ANKRD13A Rabbit Polyclonal Antibody

Supplier: Proteintech
23998-1-AP
10156-100EA 635.16 CAD
10156-100
Anti-ANKRD13A Rabbit Polyclonal Antibody
Antibodies
The ANKRD13A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ANKRD13A. This antibody recognizes human antigen. The ANKRD13A antibody has been validated for the following applications: ELISA, WB, IF analysis.

WB 1 : 1000 ; IF 1 : 50

Type: Primary
Antigen: ANKRD13A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR