Anti-IL32 Rabbit Polyclonal Antibody

Supplier: Proteintech
11079-1-AP
10155-966EA 635.16 CAD
10155-966
Anti-IL32 Rabbit Polyclonal Antibody
Antibodies
The IL32 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human IL32. This antibody recognizes human antigen. The IL32 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 800 ; IHC 1 : 50 ; IP 1 : 500 ; IF 1 : 25

Type: Primary
Antigen: IL32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR