Anti-PCGF6 Rabbit Polyclonal Antibody

Supplier: Proteintech
24103-1-AP-150UL
10155-146EA 635.16 CAD
10155-146
Anti-PCGF6 Rabbit Polyclonal Antibody
Antibodies
The PCGF6 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PCGF6. This antibody recognizes human antigen. The PCGF6 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 500

Type: Primary
Antigen: PCGF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR