Anti-GCK Rabbit Polyclonal Antibody

Supplier: Proteintech
15629-1-AP-150UL
10154-622EA 635.16 CAD
10154-622
Anti-GCK Rabbit Polyclonal Antibody
Antibodies
The GCK antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human GCK. This antibody recognizes human,mouse,rat antigen. The GCK antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 2000 ; IHC 1 : 100

Type: Primary
Antigen: GCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR