Anti-A4GALT Rabbit Polyclonal Antibody

Supplier: Proteintech
12392-1-AP-150UL
10154-484EA 635.16 CAD
10154-484
Anti-A4GALT Rabbit Polyclonal Antibody
Antibodies
The A4GALT antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human A4GALT. This antibody recognizes human, mouse antigen. The A4GALT antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; FC 1 : 0

Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR