Anti-ISM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21540-1-AP
10154-964EA 635.16 CAD
10154-964
Anti-ISM2 Rabbit Polyclonal Antibody
Antibodies
The ISM2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ISM2. This antibody recognizes human, mouse antigen. The ISM2 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: ISM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR