Anti-JMJD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21572-1-AP
10154-968EA 635.16 CAD
10154-968
Anti-JMJD4 Rabbit Polyclonal Antibody
Antibodies
The JMJD4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human JMJD4. This antibody recognizes human antigen. The JMJD4 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: JMJD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR