Anti-WDR16 Rabbit Polyclonal Antibody

Supplier: Proteintech
12644-1-AP
10155-996EA 635.16 CAD
10155-996
Anti-WDR16 Rabbit Polyclonal Antibody
Antibodies
The WDR16 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human WDR16. This antibody recognizes human,mouse,rat antigen. The WDR16 antibody has been validated for the following applications: ELISA, IHC analysis.

IHC 1 : 50 ; WB 1 : 200

Type: Primary
Antigen: WDR16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR