Anti-MNS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12693-1-AP
10154-488EA 635.16 CAD
10154-488
Anti-MNS1 Rabbit Polyclonal Antibody
Antibodies
The MNS1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MNS1. This antibody recognizes human,mouse,rat antigen. The MNS1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

FC 1 : 0 ; IP 1 : 1000

Type: Primary
Antigen: MNS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR