Anti-Integrin beta-6 Rabbit Polyclonal Antibody

Supplier: Proteintech
21703-1-AP
10154-982EA 635.16 CAD
10154-982
Anti-Integrin beta-6 Rabbit Polyclonal Antibody
Antibodies
The Integrin beta-6 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Integrin beta-6. This antibody recognizes human,mouse,rat antigen. The Integrin beta-6 antibody has been validated for the following applications: ELISA, IHC analysis.

IHC 1 : 50

Type: Primary
Antigen: Integrin beta-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR