Anti-GAD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21760-1-AP
10154-986EA 635.16 CAD
10154-986
Anti-GAD2 Rabbit Polyclonal Antibody
Antibodies
The GAD2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human GAD2. This antibody recognizes human, mouse, rat antigen. The GAD2 antibody has been validated for the following applications: ELISA, WB, IP analysis.

WB 1 : 1000

Type: Primary
Antigen: GAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR