Anti-C11ORF54 Rabbit Polyclonal Antibody

Supplier: Proteintech
23251-1-AP-150UL
10155-106EA 635.16 CAD
10155-106
Anti-C11ORF54 Rabbit Polyclonal Antibody
Antibodies
The C11orf54 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C11orf54. This antibody recognizes human, rat antigen. The C11orf54 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: C11ORF54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR