Anti-MASP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21837-1-AP-150UL
10154-990EA 635.16 CAD
10154-990
Anti-MASP1 Rabbit Polyclonal Antibody
Antibodies
The MASP1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MASP1. This antibody recognizes human antigen. The MASP1 antibody has been validated for the following applications: ELISA, WB analysis.

LL

Type: Primary
Antigen: MASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR