Anti-CGRRF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16101-1-AP
10154-652EA 635.16 CAD
10154-652
Anti-CGRRF1 Rabbit Polyclonal Antibody
Antibodies
The CGRRF1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CGRRF1. This antibody recognizes human, rat antigen. The CGRRF1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IF 1 : 50 ; IHC 1 : 50

Type: Primary
Antigen: CGRRF1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR