Anti-IGHG4 Rabbit Polyclonal Antibody

Supplier: Proteintech
16402-1-AP
10154-662EA 635.16 CAD
10154-662
Anti-IGHG4 Rabbit Polyclonal Antibody
Antibodies
The IGHG4 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human IGHG4. This antibody recognizes human,mouse,rat antigen. The IGHG4 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: IGHG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR