Anti-TEKT3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12959-1-AP-150UL
10155-998EA 635.16 CAD
10155-998
Anti-TEKT3 Rabbit Polyclonal Antibody
Antibodies
The TEKT3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TEKT3. This antibody recognizes human,mouse,rat antigen. The TEKT3 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: TEKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR