Anti-ASIC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17851-1-AP
10154-770EA 635.16 CAD
10154-770
Anti-ASIC2 Rabbit Polyclonal Antibody
Antibodies
The ASIC2 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human ASIC2. This Antibody recognizes human,mouse,rat Antigen. The ASIC2 Antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: ASIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR