Anti-FBXW5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18321-1-AP
10154-812EA 635.16 CAD
10154-812
Anti-FBXW5 Rabbit Polyclonal Antibody
Antibodies
The FBXW5 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FBXW5. This antibody recognizes human,mouse,rat antigen. The FBXW5 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: FBXW5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR