Anti-CD24 Rabbit Polyclonal Antibody

Supplier: Proteintech
18330-1-AP
10154-814EA 635.16 CAD
10154-814
Anti-CD24 Rabbit Polyclonal Antibody
Antibodies
The CD24 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CD24. This antibody recognizes human, mouse antigen. The CD24 antibody has been validated for the following applications: ELISA, WB analysis.

LL

Type: Primary
Antigen: CD24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR