Anti-UNC5CL Rabbit Polyclonal Antibody

Supplier: Proteintech
13322-1-AP
10154-514EA 635.16 CAD
10154-514
Anti-UNC5CL Rabbit Polyclonal Antibody
Antibodies
The UNC5CL antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human UNC5CL. This antibody recognizes human, mouse antigen. The UNC5CL antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 1000

Type: Primary
Antigen: UNC5CL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR