Anti-RRBP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22015-1-AP
10155-014EA 818.44 CAD
10155-014
Anti-RRBP1 Rabbit Polyclonal Antibody
Antibodies
The RRBP1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RRBP1. This antibody recognizes human, mouse antigen. The RRBP1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: RRBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR