Anti-PPP2R2A Rabbit Polyclonal Antibody

Supplier: Proteintech
16569-1-AP-150UL
10154-676EA 635.16 CAD
10154-676
Anti-PPP2R2A Rabbit Polyclonal Antibody
Antibodies
The PPP2R2A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PPP2R2A. This antibody recognizes human,mouse,rat antigen. The PPP2R2A antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: PPP2R2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR