Chromegabond® High Carbon ODS C18, Traditional Reverse Phase HPLC Columns, 100 Å, ES Industries
Supplier: Avantor
155221HCC18
95001-248EA 1372.37 CAD
95001-248 95001-250 95001-254 95001-256 95001-258 95001-262 95001-264 95001-266 95001-270 95001-272 95001-274 95001-278 95001-280 95001-282 95001-286 95001-288 95001-290 95001-294 95001-296 95001-298
Chromegabond® High Carbon ODS C18, Traditional Reverse Phase HPLC Columns, 100 Å, ES Industries
Chromatography Columns
ES Industries has developed a wide range of Chromegabond reverse phase columns to provide the means of separating a broad range of compounds.
  • Provide superior, reproducible separations
  • Long column life with excellent peak symmetry
These phases are manufactured using established procedures and have been produced for a number of years to provide the chromatographer/QC chemist with continuous stream of highly reproducible columns.