Anti-GP repeat Rabbit Polyclonal Antibody

Supplier: Proteintech
24494-1-AP
10155-214EA 828.39 CAD
10155-214
Anti-GP repeat Rabbit Polyclonal Antibody
Antibodies
The GP repeat antibody from Proteintech is a rabbit polyclonal antibody to a peptide. This antibody recognizes human antigen. The GP repeat antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: GP repeat
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR