Anti-ARID4B Rabbit Polyclonal Antibody

Supplier: Proteintech
24499-1-AP-150UL
10155-218EA 635.16 CAD
10155-218
Anti-ARID4B Rabbit Polyclonal Antibody
Antibodies
The ARID4B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ARID4B. This antibody recognizes human antigen. The ARID4B antibody has been validated for the following applications: ELISA, WB, IF, IHC, IP analysis.

WB 1 : 1000

Type: Primary
Antigen: ARID4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR