Anti-POTEA Rabbit Polyclonal Antibody

Supplier: Proteintech
24593-1-AP
10155-248EA 635.16 CAD
10155-248
Anti-POTEA Rabbit Polyclonal Antibody
Antibodies
The POTEA antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human POTEA. This antibody recognizes human, mouse antigen. The POTEA antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: POTEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR