Anti-MBNL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
24610-1-AP
10155-258EA 635.16 CAD
10155-258
Anti-MBNL3 Rabbit Polyclonal Antibody
Antibodies
The MBNL3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MBNL3. This antibody recognizes human, mouse antigen. The MBNL3 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 600 ; IP 1 : 500

Type: Primary
Antigen: MBNL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR