Anti-ZBTB48 Rabbit Polyclonal Antibody

Supplier: Proteintech
24665-1-AP
10155-272EA 635.16 CAD
10155-272
Anti-ZBTB48 Rabbit Polyclonal Antibody
Antibodies
The ZBTB48 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZBTB48. This antibody recognizes human antigen. The ZBTB48 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300

Type: Primary
Antigen: ZBTB48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR