Anti-SAMD11 Rabbit Polyclonal Antibody

Supplier: Proteintech
24731-1-AP
10155-292EA 635.16 CAD
10155-292
Anti-SAMD11 Rabbit Polyclonal Antibody
Antibodies
The SAMD11 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SAMD11. This antibody recognizes human antigen. The SAMD11 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: SAMD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR