Anti-HYDIN Rabbit Polyclonal Antibody

Supplier: Proteintech
24741-1-AP-150UL
10155-298EA 635.16 CAD
10155-298
Anti-HYDIN Rabbit Polyclonal Antibody
Antibodies
The HYDIN antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human HYDIN. This antibody recognizes human antigen. The HYDIN antibody has been validated for the following applications: ELISA, IHC, WB analysis.

IHC 1 : 50 ; IP 1 : 500

Type: Primary
Antigen: HYDIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR