Anti-IDNK Rabbit Polyclonal Antibody

Supplier: Proteintech
24831-1-AP-150UL
10155-328EA 635.16 CAD
10155-328
Anti-IDNK Rabbit Polyclonal Antibody
Antibodies
The IDNK antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human IDNK. This antibody recognizes human, mouse antigen. The IDNK antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: IDNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR