Anti-SEC61A Rabbit Polyclonal Antibody

Supplier: Proteintech
24935-1-AP-150UL
10155-364EA 635.16 CAD
10155-364
Anti-SEC61A Rabbit Polyclonal Antibody
Antibodies
The SEC61alpha antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SEC61alpha. This antibody recognizes human, mouse antigen. The SEC61alpha antibody has been validated for the following applications: ELISA, WB, IF analysis.

WB 1 : 700 ; IHC 1 : 50

Type: Primary
Antigen: SEC61A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR