Anti-C6orf15 Rabbit Polyclonal Antibody

Supplier: Proteintech
24953-1-AP
10155-374EA 635.16 CAD
10155-374
Anti-C6orf15 Rabbit Polyclonal Antibody
Antibodies
The C6orf15 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C6orf15. This antibody recognizes human, monkey antigen. The C6orf15 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 50 ; WB 1 : 300

Type: Primary
Antigen: C6orf15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR