Anti-LCE1A Rabbit Polyclonal Antibody

Supplier: Proteintech
24956-1-AP-150UL
10155-376EA 635.16 CAD
10155-376
Anti-LCE1A Rabbit Polyclonal Antibody
Antibodies
The LCE1A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LCE1A. This antibody recognizes human antigen. The LCE1A antibody has been validated for the following applications: ELISA, IHC analysis.

IHC 1 : 50

Type: Primary
Antigen: LCE1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR