Anti-FDXACB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24981-1-AP
10155-386EA 635.16 CAD
10155-386
Anti-FDXACB1 Rabbit Polyclonal Antibody
Antibodies
The FDXACB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FDXACB1. This antibody recognizes human antigen. The FDXACB1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: FDXACB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR