Anti-C9orf61 Rabbit Polyclonal Antibody

Supplier: Proteintech
25630-1-AP
10155-736EA 635.16 CAD
10155-736
Anti-C9orf61 Rabbit Polyclonal Antibody
Antibodies
The C9orf61 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C9orf61. This antibody recognizes human, mouse antigen. The C9orf61 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: C9orf61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR