Anti-SCGB2A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25645-1-AP-150UL
10156-306EA 635.16 CAD
10156-306
Anti-SCGB2A2 Rabbit Polyclonal Antibody
Antibodies
The Mammaglobin A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Mammaglobin A. This antibody recognizes human antigen. The Mammaglobin A antibody has been validated for the following applications: ELISA, IHC analysis.

IP 1 : 1000 ; WB 1 : 1500

Type: Primary
Antigen: SCGB2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR