Anti-RELB Rabbit Polyclonal Antibody

Supplier: Proteintech
25027-1-AP
10156-114EA 635.16 CAD
10156-114
Anti-RELB Rabbit Polyclonal Antibody
Antibodies
The RELB antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RELB. This antibody recognizes human antigen. The RELB antibody has been validated for the following applications: ELISA, IF, IHC, IP, WB analysis.

WB 1 : 300

Type: Primary
Antigen: RELB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR