Anti-C4orf29 Rabbit Polyclonal Antibody

Supplier: Proteintech
25036-1-AP
10155-396EA 635.16 CAD
10155-396
Anti-C4orf29 Rabbit Polyclonal Antibody
Antibodies
The C4orf29 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C4orf29. This antibody recognizes human, mouse antigen. The C4orf29 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

WB 1 : 600

Type: Primary
Antigen: C4orf29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR